siRNA Details
| Gene Symbol: Gm15972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090376.1 | - | 1 | 1821 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY755527.1 | - | 1 | 658 | Chr. 18 - 23750456 - 23751160 [-] on Build GRCm38 | |

Gm15972
Genomic Map