siRNA Details
| Gene Symbol: Gm11269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132790.1 | - | 3 | 368 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614867.1 | - | 3 | 365 | Chr. 4 - 82692805 - 82693711 [+] on Build GRCm38 | |

Gm11269
Genomic Map