siRNA Details
| Gene Symbol: E130218I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040435.1 | 5 | 672 | |||
| NR_040436.1 | 5 | 627 | ||||
| GenBank mRNA | AK021371.1 | - | 5 | 628 | ||
| AK087464.1 | - | 3 | 1403 | |||
| BC083116.1 | - | 4 | 934 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130218I03Rik| Gene Symbol: E130218I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040435.1 | 5 | 672 | |||
| NR_040436.1 | 5 | 627 | ||||
| GenBank mRNA | AK021371.1 | - | 5 | 628 | ||
| AK087464.1 | - | 3 | 1403 | |||
| BC083116.1 | - | 4 | 934 | |||