siRNA Details
| Gene Symbol: Slfn5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045932.1 | 3 | 538 | |||
| GenBank mRNA | AK005634.1 | - | 3 | 515 | ||
| BY705829.1 | - | 3 | 515 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464433.1 | - | 3 | 541 | Chr. 11 - 82942306 - 82960681 [-] on Build GRCm38 | |

Slfn5os
Genomic Map