siRNA Details
| Gene Symbol: Slfn5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045932.1 | 3 | 559 | |||
| GenBank mRNA | AK005634.1 | - | 3 | 536 | ||
| BY705829.1 | - | 3 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn5os| Gene Symbol: Slfn5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045932.1 | 3 | 559 | |||
| GenBank mRNA | AK005634.1 | - | 3 | 536 | ||
| BY705829.1 | - | 3 | 536 | |||