siRNA Details
| Gene Symbol: 1700017J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040326.1 | 2 | 307 | |||
| GenBank mRNA | AK006070.1 | - | 2 | 308 | ||
| BC048565.1 | - | 2 | 307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700017J07Rik| Gene Symbol: 1700017J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040326.1 | 2 | 307 | |||
| GenBank mRNA | AK006070.1 | - | 2 | 308 | ||
| BC048565.1 | - | 2 | 307 | |||