siRNA Details
Gene Symbol: 1700017J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040326.1 | 1 | 207 | |||
GenBank mRNA | AK006070.1 | - | 1 | 208 | ||
BC048565.1 | - | 1 | 207 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700017J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040326.1 | 1 | 207 | |||
GenBank mRNA | AK006070.1 | - | 1 | 208 | ||
BC048565.1 | - | 1 | 207 |