siRNA Details
| Gene Symbol: 4930593C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040753.1 | 3 | 499 | |||
| GenBank mRNA | AK019838.1 | - | 3 | 504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930593C16Rik| Gene Symbol: 4930593C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040753.1 | 3 | 499 | |||
| GenBank mRNA | AK019838.1 | - | 3 | 504 | ||