siRNA Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_028380.1 | 4 | 505 | |||
| NR_028381.1 | 4 | 526 | ||||
| GenBank mRNA | AK020989.1 | - | LIT1001 Ftx |
4 | 393 | |
| AK035525.1 | - | 4 | 528 | |||
| AK085125.1 | - | 4 | 392 | |||
| BC027389.1 | - | 3 | 301 | |||
| BC076616.1 | - | 4 | 414 | |||

Ftx
Genomic Map