siRNA Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028380.1 | 10 | 1253 | |||
| NR_028381.1 | 9 | 1112 | ||||
| GenBank mRNA | AK035525.1 | - | 9 | 1114 | ||
| AK085125.1 | - | 10 | 1141 | |||
| BC027389.1 | - | 9 | 1049 | |||
| BC076616.1 | - | 10 | 1162 | |||

Ftx
Genomic Map