siRNA Details
| Gene Symbol: Gm14123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076437.1 | - | 3 | 2442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14123| Gene Symbol: Gm14123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076437.1 | - | 3 | 2442 | ||