siRNA Details
| Gene Symbol: Gt(ROSA)26Sor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_027008.1 | 2 | 680 | |||
| NR_027009.1 | 2 | 574 | ||||
| GenBank mRNA | U83174.1 | - | LIT3416 Rosa26.1 |
2 | 633 | |

Gt(ROSA)26Sor
Genomic Map