siRNA Details
| Gene Symbol: 4930551O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016096.1 | - | 2 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930551O13Rik| Gene Symbol: 4930551O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016096.1 | - | 2 | 293 | ||