siRNA Details
| Gene Symbol: Gm12069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158601.1 | - | 3 | 325 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY240988.1 | - | 3 | 323 | Chr. 11 - 25301263 - 25322448 [+] on Build GRCm38 | |

Gm12069
Genomic Map