siRNA Details
| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040684.1 | 5 | 695 | |||
| GenBank mRNA | AK016926.1 | - | 5 | 694 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716606.1 | - | 691 | Chr. - - [-] on Build | ||

Prkag2os1
Genomic Map