siRNA Details
| Gene Symbol: 9030204H09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040618.1 | 3 | 566 | |||
| GenBank mRNA | AK020243.1 | - | 3 | 567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030204H09Rik| Gene Symbol: 9030204H09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040618.1 | 3 | 566 | |||
| GenBank mRNA | AK020243.1 | - | 3 | 567 | ||