siRNA Details
| Gene Symbol: Macrod2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031201.1 | - | 3, 4 | 491 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB620502.1 | - | 3, 4 | 491 | Chr. 2 - 141249699 - 141286610 [-] on Build GRCm38 | |

Macrod2os1
Genomic Map