siRNA Details
| Gene Symbol: 4930500J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040323.1 | 2 | 679 | |||
| GenBank mRNA | CF898122.1 | - | 2 | 568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930500J02Rik| Gene Symbol: 4930500J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040323.1 | 2 | 679 | |||
| GenBank mRNA | CF898122.1 | - | 2 | 568 | ||