siRNA Details
| Gene Symbol: 1700030C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132720.1 | - | 2 | 266 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ046762.1 | - | 2 | 266 | Chr. 11 - 23497783 - 23499004 [+] on Build GRCm38 | |

1700030C12Rik
Genomic Map