siRNA Details
| Gene Symbol: Etaa1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019730.1 | - | 2 | 190 | ||
| BY715599.1 | - | 2 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etaa1os| Gene Symbol: Etaa1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019730.1 | - | 2 | 190 | ||
| BY715599.1 | - | 2 | 190 | |||