siRNA Details
| Gene Symbol: 2210417A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028285.1 | 1 | 31 | |||
| GenBank mRNA | AK008960.1 | - | 1 | 31 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210417A02Rik| Gene Symbol: 2210417A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028285.1 | 1 | 31 | |||
| GenBank mRNA | AK008960.1 | - | 1 | 31 | ||