siRNA Details
| Gene Symbol: B130055M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045290.1 | - | 2 | 1589 | ||
| AK050836.1 | - | 2 | 1593 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B130055M24Rik| Gene Symbol: B130055M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045290.1 | - | 2 | 1589 | ||
| AK050836.1 | - | 2 | 1593 | |||