siRNA Details
Gene Symbol: Gm11351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045062.1 | 4 | 468 | |||
GenBank mRNA | AK132076.1 | - | 4 | 468 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045062.1 | 4 | 468 | |||
GenBank mRNA | AK132076.1 | - | 4 | 468 |