siRNA Details
| Gene Symbol: 9530034E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020586.1 | - | 3 | 317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530034E10Rik| Gene Symbol: 9530034E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020586.1 | - | 3 | 317 | ||