siRNA Details
| Gene Symbol: Rab11fip4os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045898.2 | 2, 3 | 181 | |||
| GenBank mRNA | AK019731.1 | - | 2, 3 | 183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip4os2| Gene Symbol: Rab11fip4os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045898.2 | 2, 3 | 181 | |||
| GenBank mRNA | AK019731.1 | - | 2, 3 | 183 | ||