siRNA Details
| Gene Symbol: Gm16548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037987.1 | 2 | 431 | |||
| GenBank mRNA | AK149537.1 | - | 2 | 204 | ||
| BC019478.1 | - | 2 | 431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16548| Gene Symbol: Gm16548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037987.1 | 2 | 431 | |||
| GenBank mRNA | AK149537.1 | - | 2 | 204 | ||
| BC019478.1 | - | 2 | 431 | |||