siRNA Details
| Gene Symbol: C030005K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043286.1 | - | 3 | 497 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640727.1 | - | 3 | 497 | Chr. 18 - 50052299 - 50053844 [-] on Build GRCm38 | |

C030005K06Rik
Genomic Map