siRNA Details
| Gene Symbol: 4930511M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015494.1 | 3 | 856 | |||
| NR_027944.1 | 2 | 701 | ||||
| GenBank mRNA | AK144587.1 | - | 3 | 824 | ||
| AK144776.1 | - | 2 | 702 | |||
| AK165876.1 | - | 3 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930511M06Rik| Gene Symbol: 4930511M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015494.1 | 3 | 856 | |||
| NR_027944.1 | 2 | 701 | ||||
| GenBank mRNA | AK144587.1 | - | 3 | 824 | ||
| AK144776.1 | - | 2 | 702 | |||
| AK165876.1 | - | 3 | 857 | |||