siRNA Details
| Gene Symbol: Gm16157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040340.1 | 4 | 1008 | |||
| NR_040341.1 | 3 | 943 | ||||
| GenBank mRNA | AK149394.1 | - | 3 | 944 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16157| Gene Symbol: Gm16157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040340.1 | 4 | 1008 | |||
| NR_040341.1 | 3 | 943 | ||||
| GenBank mRNA | AK149394.1 | - | 3 | 944 | ||