siRNA Details
| Gene Symbol: 1700112J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077218.1 | 2 | 242 | |||
| GenBank mRNA | AK018912.1 | - | 2 | 228 | ||
| AK018948.1 | - | 2 | 243 | |||
| AK028097.1 | - | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700112J05Rik| Gene Symbol: 1700112J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077218.1 | 2 | 242 | |||
| GenBank mRNA | AK018912.1 | - | 2 | 228 | ||
| AK018948.1 | - | 2 | 243 | |||
| AK028097.1 | - | 2 | 230 | |||