siRNA Details
| Gene Symbol: 1700112J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077218.1 | 2, 3 | 284 | |||
| GenBank mRNA | AK018948.1 | - | 2, 3 | 285 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707240.1 | - | 2, 3 | 285 | Chr. 5 - 77006508 - 77018863 [-] on Build GRCm38 | |

1700112J05Rik
Genomic Map