siRNA Details
| Gene Symbol: Gm16876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040110.1 | - | 279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16876| Gene Symbol: Gm16876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040110.1 | - | 279 | |||