siRNA Details
Gene Symbol: Gm15247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019053.1 | - | 2 | 378 | ||
AV076505.1 | - | 2 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708654.1 | - | 2 | 377 | Chr. X - 169985801 - 169986940 [-] on Build GRCm38 |
