siRNA Details
| Gene Symbol: E130102H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040708.1 | 2 | 601 | |||
| GenBank mRNA | AK021368.1 | - | 2 | 563 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY731240.1 | - | 2 | 563 | Chr. 4 - 101346569 - 101356209 [-] on Build GRCm38 | |

E130102H24Rik
Genomic Map