siRNA Details
| Gene Symbol: Gm11482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077143.1 | - | 4 | 550 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617036.1 | - | 4 | 549 | Chr. 4 - 63634421 - 63639361 [+] on Build GRCm38 | |

Gm11482
Genomic Map