siRNA Details
| Gene Symbol: Ctrcos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040641.1 | 3 | 271 | |||
| GenBank mRNA | AK007697.1 | - | 3 | 272 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV055468.1 | - | 2 | 228 | Chr. 4 - 141846512 - 141846990 [+] on Build GRCm38 | |

Ctrcos
Genomic Map