siRNA Details
| Gene Symbol: Gm13748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY214029.1 | - | 112 | |||
| BY222622.1 | - | 111 | ||||
| BY223622.1 | - | 111 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13748| Gene Symbol: Gm13748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY214029.1 | - | 112 | |||
| BY222622.1 | - | 111 | ||||
| BY223622.1 | - | 111 | ||||