siRNA Details
| Gene Symbol: Gm1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046015.1 | 8 | 647 | |||
| GenBank mRNA | AK144520.1 | - | 8 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1720| Gene Symbol: Gm1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046015.1 | 8 | 647 | |||
| GenBank mRNA | AK144520.1 | - | 8 | 648 | ||