siRNA Details
| Gene Symbol: Gm13546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045895.1 | 4 | 968 | |||
| NR_045896.1 | 2 | 635 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13546| Gene Symbol: Gm13546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045895.1 | 4 | 968 | |||
| NR_045896.1 | 2 | 635 | ||||