siRNA Details
| Gene Symbol: 4933430M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045857.2 | 4 | 421 | |||
| GenBank mRNA | AK016999.1 | - | 4 | 422 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716684.1 | - | 4 | 422 | Chr. 11 - 24481909 - 24569952 [+] on Build GRCm38 | |

4933430M04Rik
Genomic Map