siRNA Details
Gene Symbol: Frs3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045912.1 | 3 | 645 | |||
NR_045913.1 | 3 | 610 | ||||
GenBank mRNA | AK076701.1 | - | 3 | 646 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Frs3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045912.1 | 3 | 645 | |||
NR_045913.1 | 3 | 610 | ||||
GenBank mRNA | AK076701.1 | - | 3 | 646 |