siRNA Details
| Gene Symbol: Gm13066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC106196.1 | - | 1 | 125 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ930211.1 | - | 1 | 118 | Chr. 4 - 149565678 - 149567790 [-] on Build GRCm38 | |

Gm13066
Genomic Map