siRNA Details
| Gene Symbol: 1700051A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045922.1 | 2 | 1299 | |||
| GenBank mRNA | AK035540.1 | - | 2 | 1299 | ||
| AK077134.1 | - | 3 | 1440 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700051A21Rik| Gene Symbol: 1700051A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045922.1 | 2 | 1299 | |||
| GenBank mRNA | AK035540.1 | - | 2 | 1299 | ||
| AK077134.1 | - | 3 | 1440 | |||