siRNA Details
| Gene Symbol: 4933431E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015459.1 | 1 | 515 | |||
| GenBank mRNA | AK030251.1 | - | 1 | 70 | ||
| AK036227.1 | - | 1 | 101 | |||
| AK043835.1 | - | 1 | 76 | |||
| AK147824.1 | - | 1 | 44 | |||
| BC060136.1 | - | 1 | 515 | |||

4933431E20Rik
Genomic Map