siRNA Details
| Gene Symbol: 4930443O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040504.1 | 3 | 328 | |||
| GenBank mRNA | AK015373.1 | - | 3 | 329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930443O20Rik| Gene Symbol: 4930443O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040504.1 | 3 | 328 | |||
| GenBank mRNA | AK015373.1 | - | 3 | 329 | ||