siRNA Details
| Gene Symbol: Gm13544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040365.1 | 4 | 697 | |||
| GenBank mRNA | AK144626.1 | - | 4 | 698 | ||
| AK165961.1 | - | 4 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13544| Gene Symbol: Gm13544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040365.1 | 4 | 697 | |||
| GenBank mRNA | AK144626.1 | - | 4 | 698 | ||
| AK165961.1 | - | 4 | 697 | |||