siRNA Details
| Gene Symbol: B230119M05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045454.1 | 3 | 935 | |||
| GenBank mRNA | AK020983.1 | - | 3 | 936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230119M05Rik| Gene Symbol: B230119M05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045454.1 | 3 | 935 | |||
| GenBank mRNA | AK020983.1 | - | 3 | 936 | ||