siRNA Details
| Gene Symbol: Gm12505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040674.1 | 2 | 190 | |||
| GenBank mRNA | BC049732.1 | - | 190 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA463700.1 | - | 2 | 193 | Chr. 4 - 55410443 - 55418845 [-] on Build GRCm38 | |

Gm12505
Genomic Map