siRNA Details
| Gene Symbol: Tmem132cos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038127.1 | 3 | 394 | |||
| NR_038128.1 | 4 | 462 | ||||
| NR_038129.1 | 4 | 604 | ||||
| GenBank mRNA | AK007002.1 | - | 3 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132cos| Gene Symbol: Tmem132cos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038127.1 | 3 | 394 | |||
| NR_038128.1 | 4 | 462 | ||||
| NR_038129.1 | 4 | 604 | ||||
| GenBank mRNA | AK007002.1 | - | 3 | 357 | ||