siRNA Details
| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038037.1 | 2 | 320 | |||
| GenBank mRNA | AK087502.1 | - | 2 | 191 | ||
| AK164126.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130307A14Rik| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038037.1 | 2 | 320 | |||
| GenBank mRNA | AK087502.1 | - | 2 | 191 | ||
| AK164126.1 | - | 2 | 280 | |||